Learn More
Cayman Chemical (+)-γ-Tocopherol, Purity: ≥90%, CAS: 54-28-4, 50 mg
SDP
Catalog No. 502912243
Encompass_Preferred
A biologically active form of vitamin E; inhibits the growth of DU145, LNCaP, and Caco-2 cancer cells at 25 µM; reduces airway eosinophil infiltration and levels of IL-6, IL-5, IL-13, and CCL2 in a rat model of ovalbumin-induced allergic inflammation and a rat model of ovalbumin- and ozone-induced asthma at 100 mg/kg; reduces lipid peroxidation in vitamin E-depleted, iron-loaded rats
Catalog No. 50-291-2243 Supplier Cayman Chemical Supplier No. 3636250 MG
A biologically active form of vitamin E; inhibits the growth of DU145, LNCaP, and Caco-2 cancer cells at 25 µM; reduces airway eosinophil infiltration and levels of IL-6, IL-5, IL-13, and CCL2 in a rat model of ovalbumin-induced allergic inflammation and a rat model of ovalbumin- and ozone-induced asthma at 100 mg/kg; reduces lipid peroxidation in vitamin E-depleted, iron-loaded rats