Learn More
Medchemexpress LLC γ-Tocopherol | 54-28-4 | 25 MG
SDP
Catalog No. 5000422003
Encompass_Preferred
γ-Tocopherol is a chemical compound primarily intended for use as a laboratory chemical and in the manufacture of substances. This product is for research use only and has not been fully validated for medical applications. It is stable under recommended storage conditions.
  • Intended for laboratory use.
  • Stable under recommended storage conditions.
  • Recommended storage at 4°C, protected from light.
  • For shipping, can be at room temperature if less than 2 weeks.
  • Keep container tightly sealed in a cool, well-ventilated area.
  • Keep away from direct sunlight and sources of ignition.
Catalog No. 50-004-22003 Supplier Medchemexpress LLC Supplier No. 25MGHYN7148
γ-Tocopherol is a chemical compound primarily intended for use as a laboratory chemical and in the manufacture of substances. This product is for research use only and has not been fully validated for medical applications. It is stable under recommended storage conditions.
  • Intended for laboratory use.
  • Stable under recommended storage conditions.
  • Recommended storage at 4°C, protected from light.
  • For shipping, can be at room temperature if less than 2 weeks.
  • Keep container tightly sealed in a cool, well-ventilated area.
  • Keep away from direct sunlight and sources of ignition.