Learn More
Medchemexpress LLC (±)-α-Tocopherol acetate | 7695-91-2 | 98.7% | 100 MG
SDP
Catalog No. 5000412961
Encompass_Preferred
(±)-α-Tocopherol acetate is an orally active synthetic form of vitamin E and an ester of acetic acid and α-tocopherol. It is used in research to study the susceptibility of farmed fish to infectious diseases. In vivo studies showed that oral administration of 1200 mg/kg for 30 days stimulated the seabream's non-specific immune system and enhanced serum haemolytic activity and phagocytosis of head-kidney leucocytes.
  • Orally active synthetic form of vitamin E.
  • Ester of acetic acid and α-tocopherol.
  • Can be used for research on the susceptibility of farmed fish to infectious diseases.
  • Appears as a colorless to light yellow viscous liquid.
Catalog No. 50-004-12961 Supplier Medchemexpress LLC Supplier No. HYB1278B100MG
(±)-α-Tocopherol acetate is an orally active synthetic form of vitamin E and an ester of acetic acid and α-tocopherol. It is used in research to study the susceptibility of farmed fish to infectious diseases. In vivo studies showed that oral administration of 1200 mg/kg for 30 days stimulated the seabream's non-specific immune system and enhanced serum haemolytic activity and phagocytosis of head-kidney leucocytes.
  • Orally active synthetic form of vitamin E.
  • Ester of acetic acid and α-tocopherol.
  • Can be used for research on the susceptibility of farmed fish to infectious diseases.
  • Appears as a colorless to light yellow viscous liquid.