Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STA PHARMACEUTICAL US LLC Tocopherol amidite | 1 g | CAS 142386-74-1 | InChIKey USUSEQXJHLFSON-UHFFFAOYSA-N

Catalog No. 502354954
Encompass

Tocopherol amidite is a Amidite reagent (Subcategory: Specialty Linker/End Modifier) sold by WuXi TIDES. Offered in 1 g. Store at -20 °C. Ships on dry ice. SDS available for reference.

Specifications
- CAS: 142386-74-1
- MDL: No data
- InChIKey: USUSEQXJHLFSON-UHFFFAOYSA-N
- Molecular Weight: 630.938761998
- Molecular Formula: C38H67N2O3P
- Purity: ≥95%
- Container Type: 15 mL HDPE
- Pack Size: 1 g
- Net Weight: 1 g
- Gross Weight: 7.8 g
- Commodity Code: 29329900
- Country Of Origin: China
- IUPAC: 2-cyanoethyl (2,5,7,8-tetramethyl-2-(4,8,12-trimethyltridecyl)chroman-6-yl) diisopropylphosphoramidite
- SMILES: CC(CCCC(CCCC(CCCC1(CCC2=C(C(C)=C(C(OP(N(C(C)C)C(C)C)OCCC#N)=C2C)C)O1)C)C)C)C

Catalog No. 50-235-4954 Supplier STA PHARMACEUTICAL US LLC Supplier No. STAAM100411GM
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Tocopherol amidite is a Amidite reagent (Subcategory: Specialty Linker/End Modifier) sold by WuXi TIDES. Offered in 1 g. Store at -20 °C. Ships on dry ice. SDS available for reference.

Specifications
- CAS: 142386-74-1
- MDL: No data
- InChIKey: USUSEQXJHLFSON-UHFFFAOYSA-N
- Molecular Weight: 630.938761998
- Molecular Formula: C38H67N2O3P
- Purity: ≥95%
- Container Type: 15 mL HDPE
- Pack Size: 1 g
- Net Weight: 1 g
- Gross Weight: 7.8 g
- Commodity Code: 29329900
- Country Of Origin: China
- IUPAC: 2-cyanoethyl (2,5,7,8-tetramethyl-2-(4,8,12-trimethyltridecyl)chroman-6-yl) diisopropylphosphoramidite
- SMILES: CC(CCCC(CCCC(CCCC1(CCC2=C(C(C)=C(C(OP(N(C(C)C)C(C)C)OCCC#N)=C2C)C)O1)C)C)C)C

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.