Learn More
Medchemexpress LLC γ-Tocopherol-d4 | 1329652-13-2 | 1 MG
SDP
Catalog No. 5000421027
Encompass_Preferred
γ-Tocopherol-d4 is a deuterium-labeled form of γ-Tocopherol. The unlabeled compound, γ-Tocopherol (D-γ-Tocopherol), is recognized as a potent inhibitor of cyclooxygenase (COX) and is a naturally occurring form of Vitamin E. It is found in various plant seeds, including corn oil and soybeans. This compound is known for its anti-inflammatory properties and its anti-cancer activity.
  • Deuterium-labeled compound.
  • Acts as a potent cyclooxygenase (COX) inhibitor.
  • Is a naturally occurring form of vitamin E.
  • Demonstrates anti-inflammatory effects.
  • Exhibits anti-cancer activity.
  • Utilized as a tracer in research.
  • Serves as an internal standard for quantitative analysis using techniques such as NMR, GC-MS, or LC-MS.
  • Deuterium substitution can influence the pharmacokinetic and metabolic profiles of pharmaceutical compounds.
Catalog No. 50-004-21027 Supplier Medchemexpress LLC Supplier No. HYN7148S11MG
γ-Tocopherol-d4 is a deuterium-labeled form of γ-Tocopherol. The unlabeled compound, γ-Tocopherol (D-γ-Tocopherol), is recognized as a potent inhibitor of cyclooxygenase (COX) and is a naturally occurring form of Vitamin E. It is found in various plant seeds, including corn oil and soybeans. This compound is known for its anti-inflammatory properties and its anti-cancer activity.
  • Deuterium-labeled compound.
  • Acts as a potent cyclooxygenase (COX) inhibitor.
  • Is a naturally occurring form of vitamin E.
  • Demonstrates anti-inflammatory effects.
  • Exhibits anti-cancer activity.
  • Utilized as a tracer in research.
  • Serves as an internal standard for quantitative analysis using techniques such as NMR, GC-MS, or LC-MS.
  • Deuterium substitution can influence the pharmacokinetic and metabolic profiles of pharmaceutical compounds.