Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Tocopherols | 1406-66-2 | 98.6% | 100 MG
SDP

Catalog No. 5000435817
Encompass_Preferred

Tocopherols are an important class of fat-soluble vitamins, with gamma-tocopherol being the most common form, followed by beta-tocopherol and alpha-tocopherol. This product is for research use only and is not sold to patients.

  • Appearance: Liquid
  • Color: Colorless to light yellow
  • Store pure form at -20°C for 3 years
  • Store in solvent at -80°C for 6 months or -20°C for 1 month
  • Soluble in DMSO at 100 mg/mL
  • Classified as a natural product, reference standard, and endogenous metabolite

Catalog No. 50-004-35817 Supplier Medchemexpress LLC Supplier No. HY126400100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Tocopherols are an important class of fat-soluble vitamins, with gamma-tocopherol being the most common form, followed by beta-tocopherol and alpha-tocopherol. This product is for research use only and is not sold to patients.

  • Appearance: Liquid
  • Color: Colorless to light yellow
  • Store pure form at -20°C for 3 years
  • Store in solvent at -80°C for 6 months or -20°C for 1 month
  • Soluble in DMSO at 100 mg/mL
  • Classified as a natural product, reference standard, and endogenous metabolite

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.