Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocopheryl Acid Succinate, a Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111014201 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-4201 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-4201 Supplier MilliporeSigma Supelco Supplier No. PHR1029500MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Certified pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to pharmacopeia primary standards. Tocopheryl Acid Succinate, a is a derivative of vitamin E and a potent and highly specific anticancer agent known to inhibit the growth of cancer cells.
TRUSTED_SUSTAINABILITY

Specifications

CAS 4345-03-3
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.
Physical Form Neat
Linear Formula C33H54O5
MDL Number MFCD00072055
Synonym D-alpha-Tocopherol succinate; Vitamin E succinate
Type Certified Reference Material
Quantity 500 mg
Formula Weight 530.78
Grade Certified Reference Material
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.