Learn More
Cayman Chemical β-Tocotrienol, Purity: ≥98%, CAS: 490-23-3, 25 mg
SDP
Catalog No. 502897022
Encompass_Preferred
An isomer of Vitamin E that has less potent antioxidant properties than α-tocotrienol
Catalog No. 50-289-7022 Supplier Cayman Chemical Supplier No. 900153325 MG
An isomer of Vitamin E that has less potent antioxidant properties than α-tocotrienol