Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

β-Tocotrienol, ≥95.0% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111014304 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-4304 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-4304 Supplier MilliporeSigma Supelco Supplier No. 05644100MG
Add to Cart
Add to Cart

β-Tocotrienol is a naturally occurring forms of vitamin E commonly found in cereals such as whole wheat flour.

Featured Industries: Vitamins, Nutraceuticals and Natural Products
TRUSTED_SUSTAINABILITY

Specifications

CAS 490-23-3
Physical Form Viscous Liquid
Linear Formula C28H42O2
MDL Number MFCD09264634
Synonym (R)-beta-Tocotrienol; [R-(E,E)]-3,4-Dihydro-2,5,8-trimethyl-2-(4,8,12-trimethyl-3,7,11-tridecatrienyl)-2 H-1-benzopyran-6-ol
Type Analytical Standard
Quantity 100 mg
Formula Weight 410.63
Percent Purity ≥95.0% (HPLC)
Grade Analytical Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.