Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

δ-Tocotrienol, ≥97.0% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111013752 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-3752 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-3752 Supplier MilliporeSigma Supelco Supplier No. 69745100MG
Add to Cart
Add to Cart

δ-Tocotrienol can be found in coconut oil, palm oil, and cereal grains such as barley, oats, wheat and rye.

Featured Industries: Pharmaceutical (small molecule) Vitamins, Nutraceuticals and Natural Products
TRUSTED_SUSTAINABILITY

Specifications

CAS 25612-59-3
Physical Form Neat
Linear Formula C27H40O2
MDL Number MFCD11045309
Synonym (R)-delta-Tocotrienol; 8-Methyltocotrienol; [R-(E,E)]-3,4-Dihydro-2,8-dimethyl-2-(4,8,12-trimethyl-3,7,11-tridecatrienyl)-2 H-1-benzopyran-6-ol
Type Analytical Standard
Quantity 100 mg
Formula Weight 396.61
Percent Purity ≥97.0% (HPLC)
Grade Analytical Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.