Learn More
Tocris Bioscience™ 740 Y-P
GREENER_CHOICE
Catalog No. 19831
Change view
Click to view available options
Quantity:
1 mg
Catalog No. Quantity
19-831 1 mg
1 options
This item is not returnable. View return policy
Catalog No. 19-831 Supplier Tocris Bioscience™ Supplier No. 1983/1
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
Cell-permeable PI 3-kinase activator
740 Y-P is a cell-permeable phosphopeptide activator of PI 3-kinase. Binds with high affinity to p85 subunit of the enzyme. Displays mitogenic activity in C2 muscle cells and promotes survival of rat cerebellar granule neurons in vitro.
Specifications
Quantity 1 mg