Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Ac2-26
GREENER_CHOICE

Catalog No. 1845500U
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-455-00U 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 18-455-00U Supplier Tocris Bioscience™ Supplier No. 1845/500U
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Annexin/lipocortin 1-mimetic; inhibits leukocyte extravasation

Ac2-26 is an annexin/lipocortin 1-mimetic peptide; inhibits leukocyte extravasation. Ac2-26 reduces neutrophil adhesion and emigration, promotes detachment of neutrophils from activated mesenteric endothelium and accelerates epithelial wound repair after induced colonic injury in mice in vivo. Anti-inflammatory. Ac2-26 inhibits the proliferation of human larynx epidermoid carcinoma Hep-2 cells in vitro. Ac2-26 decreases proliferation and increases mobility of cervical cancer cells.

Specifications

Purity 95%
Content And Storage Store at -20°C
Quantity 500 μg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.