Learn More
Tocris Bioscience™ Astressin 2B
GREENER_CHOICE
Catalog No. 2391500U
Change view
Click to view available options
Quantity:
500 μg
Catalog No. Quantity
23-915-00U 500 μg
1 options
This item is not returnable. View return policy
Catalog No. 23-915-00U Supplier Tocris Bioscience™ Supplier No. 2391/500U
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
Selective CRF2 antagonist
Astressin 2B is a potent and selective corticotropin-releasing factor receptor 2 (CRF2) antagonist (IC50 values are 1.3 and > 500 nM for CRF2 and CRF1 respectively). Antagonizes CRF2-mediated inhibition of gastric emptying.
Specifications
Quantity 500 μg