Learn More
Tocris Bioscience™ Astressin
GREENER_CHOICE
Catalog No. 1606500U
Change view
Click to view available options
Quantity:
500 μg
Catalog No. Quantity
16-065-00U 500 μg
1 options
This item is not returnable. View return policy
Catalog No. 16-065-00U Supplier Tocris Bioscience™ Supplier No. 1606/500U
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
Potent CRF receptor antagonist
Astressin is a potent corticotropin-releasing factor (CRF) receptor antagonist (Ki values are 2, 1.5 and 1 nM at CRF1, CRF2α and CRF2β). Reduces ACTH secretion, blocks delayed gastric emptying and is neuroprotective in vivo.
Specifications
Quantity 500 μg