Learn More
Tocris Bioscience™ Boc-MLF
GREENER_CHOICE
Catalog No. 37301
Change view
Click to view available options
Quantity:
1 mg
Catalog No. Quantity
37-301 1 mg
1 options
This item is not returnable. View return policy
Catalog No. 37-301 Supplier Tocris Bioscience™ Supplier No. 3730/1
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
FPR1 antagonist
Boc-MLF is an antagonist of formyl peptide receptor 1 (FPR1). Reduces superoxide production induced by fMLF with an EC50 of 0.63 μM. Almost completely blocks fMLF-stimulated primary granule exocytosis.
Specifications
Quantity 1 mg