Learn More
Tocris Bioscience™ Carbetocin
GREENER_CHOICE
Catalog No. 485210
Change view
Click to view available options
Quantity:
10 mg
Catalog No. Quantity
48-521-0 10 mg
1 options
This item is not returnable. View return policy
Catalog No. 48-521-0 Supplier Tocris Bioscience™ Supplier No. 4852/10
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
Oxytocin analog
Carbetocin is an oxytocin analog. Long-acting; displays agonist properties.
Specifications
Purity 0.95
Quantity 10 mg