Learn More
Tocris Bioscience™ COG 133
GREENER_CHOICE
Catalog No. 34051
Change view
Click to view available options
Quantity:
1 mg
Catalog No. Quantity
34-051 1 mg
1 options
This item is not returnable. View return policy
Catalog No. 34-051 Supplier Tocris Bioscience™ Supplier No. 3405/1
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
ApoE peptide fragment
COG 133 is an apolipoprotein (ApoE) peptide fragment that functions via the low-density lipoprotein receptor-related protein (LRP). Substantially reduces the symptoms of experimental autoimmune encephalomyelitis, a model of human multiple sclerosis, and suppresses inflammation, demyelination and infiltration of cells into the spinal cord. Also acts as a non-competitive antagonist at α7 nicotinic acetylcholine receptors (IC50 = 445 nM).
Specifications
Purity 95%
Quantity 1 mg