Learn More
Tocris Bioscience™ [Glu27]-PKC (19-36)
Catalog No. 40591
Change view
Click to view available options
Quantity:
1 mg
Catalog No. Quantity
40-591 1 mg
1 options
This item is not returnable. View return policy
Catalog No. 40-591 Supplier Tocris Bioscience™ Supplier No. 4059/1
This item is not returnable. View return policy
Inactive control peptide for PKC (19-36) (Cat. No. 4058)
Inactive control for PKC (19-36) (Cat. No. 4058), a pseudosubstrate peptide inhibitor of protein kinase C.
Specifications
Quantity 1 mg