Learn More
Tocris Bioscience™ MOG (35-55)
GREENER_CHOICE
Catalog No. 25681
Change view
Click to view available options
Quantity:
1 mg
Catalog No. Quantity
25-681 1 mg
1 options
This item is not returnable. View return policy
Catalog No. 25-681 Supplier Tocris Bioscience™ Supplier No. 2568/1
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
Encephalitogenic myelin oligodendrocyte glycoprotein fragment
MOG (35-55) or myelin oligodendrocyte glycoprotein (MOG) 35-55 is a minor component of CNS myelin. MOG (35-55) produces a relapsing-remitting neurological disease with extensive plaque-like demyelination, common to the manifestations of multiple sclerosis. MOG (35-55) induces strong T and B cell responses and is highly encephalitogenic. MOG (35-55) induces T cell-mediated multiple sclerosis in animal models. When co-administered with ITE, MOG (35-55) induces tolerogenic dendritic cells and suppresses disease development in mouse preclinical models of multiple sclerosis.
Specifications
Quantity 1 mg