Learn More
Tocris Bioscience™ Octreotide
GREENER_CHOICE
Catalog No. 18181
Change view
Click to view available options
Quantity:
1 mg
Catalog No. Quantity
18-181 1 mg
1 options
This item is not returnable. View return policy
Catalog No. 18-181 Supplier Tocris Bioscience™ Supplier No. 1818/1
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
sst2, sst3 and sst5 agonist
Octreotide is a peptide agonist for sst2, sst3 and sst5 somatostatin receptors. IC50/Kd values (nM) at cloned human somatostatin receptors are: 290 - 1140 (sst1), 0.4 - 2.1 (sst2), 4.4 - 34.5 (sst3), > 1000 (sst4), and 5.6 - 32 (sst5).
Specifications
Quantity 1 mg