Learn More
Tocris Bioscience™ Pam2CSK4
GREENER_CHOICE
Catalog No. 46371
Change view
Click to view available options
Quantity:
1 mg
Catalog No. Quantity
46-371 1 mg
1 options
This item is not returnable. View return policy
Catalog No. 46-371 Supplier Tocris Bioscience™ Supplier No. 4637/1
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
TLR2/6 agonist; induces TNF-α production
Pam2CSK4 is a toll-like receptor 2/6 (TLR2/6) agonist. Induces TNF-α production in human mononuclear cells. Also induces proliferation and activation of mouse splenic B cells.
Specifications
Purity 95%
Quantity 1 mg