Learn More
Tocris Bioscience™ [Phe8ψ(CH-NH)-Arg9]-Bradykinin
Catalog No. 32291
Change view
Click to view available options
Quantity:
1 mg
Catalog No. Quantity
32-291 1 mg
1 options
This item is not returnable. View return policy
Catalog No. 32-291 Supplier Tocris Bioscience™ Supplier No. 3229/1
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
This item is not returnable. View return policy
Selective B2 agonist
[Phe8Ψ(CH-NH)-Arg9]-Bradykinin is a selective bradykinin B2 receptor agonist that is resistant to carboxypeptidase cleavage. 5-fold more potent and exhibits a more prolonged duration of action than bradykinin (Cat No. 3004) in vivo.
Specifications
Quantity 1 mg