Learn More
Tocris Bioscience™ Pramlintide
GREENER_CHOICE
Catalog No. 5031500U
Change view
Click to view available options
Quantity:
500 μg
Catalog No. Quantity
5031500U 500 μg
1 options
This item is not returnable. View return policy
Catalog No. 5031500U Supplier Tocris Bioscience™ Supplier No. 5031/500U
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
Synthetic version of amylin (Cat. No. 3418)
Pramlintide is a synthetic version of amylin (Cat. No. 3418). Exhibits high affinity for amylin, CGRP and calcitonin receptors (Ki values are 0.023, 3.8 and 5.1 nM respectively). Reduces postprandial hyperglycemia; also inhibits gastric emptying.
Specifications
Purity 0.95
Quantity 500 μg