Learn More
Tocris Bioscience™ Tocriscreen™ Kinase Inhibitor Toolbox I
Description
Specifications
Specifications
| Formulation | 250μL in 10mM DMSO solutions (pre- dissolved) |
| Includes | Set of FDA approved Kinase inhibitors |
| Product Type | Screening Library |
| Content And Storage | Store at -20°C |
| For Use With (Application) | Screening for therapeutic classes |
| Quantity | 80 Kinase Inhibitors |
| Format | 96-well |
For Laboratory Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.