Learn More
Tocris Bioscience™ Tocriscreen™ Kinase Inhibitor Toolbox II
Description
- Collection of 80 kinase inhibitors
- Supplied pre-dissolved in DMSO (250μL 10mM solution)
- Targets range of 40 different kinases
- Comprises subset of Tocriscreen Plus (Part. No. 5840) compound library
- Compounds: LRRK2, PI 3-kinase, mTOR, EGFR, VEGFR
- Contians novel targets: LIMK, Hapsin, NUAK1 and DYRK
- Matrix storage tubes with SepraSeal caps
Specifications
Specifications
| Formulation | 250μL in 10mM DMSO solutions (pre- dissolved) |
| Includes | Standard kinase inhibitors for targets (LRRK2, PI 3-kinase, mTOR, EGFR, VEGFR), novel targets (LIMK, Hapsin, NUAK1 and DYRK) |
| Product Type | Inhibitor |
| Content And Storage | Store at -20°C |
| For Use With (Application) | Screening for therapeutic classes |
| Quantity | 80 Kinase Inhibitors |
| Format | 96-well |
For Laboratory Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.