Learn More
Alkali Scientific Todd Hewitt Broth 100mL, 125mL Bottle, 1/Pk
SDP
Catalog No. 5000662695
Encompass_Preferred
Todd Hewitt Broth is a nutrient rich culture medium used for cultivating Streptococcus species and other fastidious bacteria. The formulation includes buffering components that prevent excessive acid buildup during bacterial growth. Streptococci produce acid as they metabolize nutrients, which can inhibit their own growth in standard media. The buffering system maintains a stable pH environment, allowing sustained growth and higher bacterial yields. This makes the medium particularly useful for clinical diagnostics, serological testing, and research applications. The 100mL bottle format provides sufficient volume for routine laboratory use and bacterial culture preparation.
Catalog No. 50-006-62695 Supplier Alkali Scientific Supplier No. THTB00
Todd Hewitt Broth is a nutrient rich culture medium used for cultivating Streptococcus species and other fastidious bacteria. The formulation includes buffering components that prevent excessive acid buildup during bacterial growth. Streptococci produce acid as they metabolize nutrients, which can inhibit their own growth in standard media. The buffering system maintains a stable pH environment, allowing sustained growth and higher bacterial yields. This makes the medium particularly useful for clinical diagnostics, serological testing, and research applications. The 100mL bottle format provides sufficient volume for routine laboratory use and bacterial culture preparation.