Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Research Products International Corp Todd Hewitt Broth, 500 Grams
SDP

Catalog No. 50489097
Encompass_Preferred

Formulated according to Updyke and Nickels modification of the medium originally described by Todd-Hewitt.

Ingredients:

Beef Heart Infusion: 3.1g/l
Dextrose: 2.0g/l
Disodium Phosphate: 0.4g/l
Yeast Enriched Peptone: 20.0g/l
Sodium Chloride: 2.0g/l
Sodium Carbonate: 2.5g/l

Catalog No. 50-489-097 Supplier Research Products International Corp Supplier No. T47500500.0
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Formulated according to Updyke and Nickels modification of the medium originally described by Todd-Hewitt.

Ingredients:

Beef Heart Infusion: 3.1g/l
Dextrose: 2.0g/l
Disodium Phosphate: 0.4g/l
Yeast Enriched Peptone: 20.0g/l
Sodium Chloride: 2.0g/l
Sodium Carbonate: 2.5g/l

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.