Learn More
NEOGEN Todd Hewitt Broth, 5 kg
SDP
Catalog No. 502015331
Encompass_Preferred
Todd Hewitt Broth is used for the cultivation of streptococci and other fastidious microorganisms. Todd Hewitt Broth is not intended for use in the diagnosis of disease or other conditions in humans. Todd Hewitt Broth was originally developed for the production of antigenic streptococcal hemolysin. Todd Hewitt Broth is prepared according to the formula described by Updyke and Nickle, who compared media for type specific extract production of group A streptococci. This study was performed using Todd Hewitt Broth prepared with infusion of fresh beef heart as a control. Results showed Todd Hewitt Broth was particularly satisfactory for growth of group A streptococci for serological typing. Todd Hewitt Broth is recommended as an enrichment medium for growth of streptococcal cells in the identification of groups A and B by IF staining.For more information visit https://www.neogen.com/solutions/Microbiology/todd-hewitt-broth
Catalog No. 50-201-5331 Supplier NEOGEN Supplier No. 700003138
Todd Hewitt Broth is used for the cultivation of streptococci and other fastidious microorganisms. Todd Hewitt Broth is not intended for use in the diagnosis of disease or other conditions in humans. Todd Hewitt Broth was originally developed for the production of antigenic streptococcal hemolysin. Todd Hewitt Broth is prepared according to the formula described by Updyke and Nickle, who compared media for type specific extract production of group A streptococci. This study was performed using Todd Hewitt Broth prepared with infusion of fresh beef heart as a control. Results showed Todd Hewitt Broth was particularly satisfactory for growth of group A streptococci for serological typing. Todd Hewitt Broth is recommended as an enrichment medium for growth of streptococcal cells in the identification of groups A and B by IF staining.For more information visit https://www.neogen.com/solutions/Microbiology/todd-hewitt-broth