Learn More
Alkali Scientific Todd Hewitt Broth with CAN 3mL 13x100, 10/Pk
SDP
Catalog No. 5000663146
Encompass_Preferred
Todd Hewitt Broth with CAN, 3mL, 13x100mm Tube, 10/Pk is a selective enrichment medium formulated for the isolation and cultivation of Group B Streptococcus (GBS) from clinical specimens, particularly in prenatal screening procedures. The broth is supplemented with the antibiotics colistin and nalidixic acid (CAN), which suppress many Gram negative bacteria and background flora while allowing selective growth of GBS organisms. Each tube contains 3mL of sterile medium in a 13x100mm screw cap tube designed for convenient inoculation and incubation in microbiology laboratories. The ready to use format supports standardized diagnostic workflows and reliable enrichment performance. This medium is commonly used in clinical diagnostics, hospital laboratories, and microbiology testing programs focused on detecting Group B Streptococcus.
Catalog No. 50-006-63146 Supplier Alkali Scientific Supplier No. THBT00
Todd Hewitt Broth with CAN, 3mL, 13x100mm Tube, 10/Pk is a selective enrichment medium formulated for the isolation and cultivation of Group B Streptococcus (GBS) from clinical specimens, particularly in prenatal screening procedures. The broth is supplemented with the antibiotics colistin and nalidixic acid (CAN), which suppress many Gram negative bacteria and background flora while allowing selective growth of GBS organisms. Each tube contains 3mL of sterile medium in a 13x100mm screw cap tube designed for convenient inoculation and incubation in microbiology laboratories. The ready to use format supports standardized diagnostic workflows and reliable enrichment performance. This medium is commonly used in clinical diagnostics, hospital laboratories, and microbiology testing programs focused on detecting Group B Streptococcus.