Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Todd Hewitt Broth with Colistin and Nalidixic Acid (LIM Broth)

Catalog No. r064810
Change view
Click to view available options
Description:
Todd Hewitt Broth; LIM Broth; 3mL; 15 x 103mm Tube
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Description
R064810 Todd Hewitt Broth; LIM Broth; 3mL; 15 x 103mm Tube
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. R064810 Supplier Thermo Scientific™ Supplier No. R064810
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Reduce media prep time while selectively isolating group B streptococci from clinical specimens containing mixed flora with pre-filled, ready-to-use tubes.

Selectively isolate group B streptococci from clinical specimens containing mixed flora using Thermo Scientific™ Todd Hewitt Broth with Colistin and Nalidixic Acid (LIM Broth).

Remove the need for pipetting, dispensing or autoclaving with convenient, pre-filled and ready-to-use tubes of Todd Hewitt Broth with the addition of Colistin and Nalidixic Acid to inhibit growth of gram-negative bacteria, available in multiple formats and formulations.

Streptococci are facultatively anaerobic, catalase-negative, gram-positive cocci that occur as pairs or chains. They cause a wide variety of clinical diseases including pharyngitis, scarlet fever, otitis media, impetigo, meningitis, neonatal sepis, pneumonia, endocarditis, osteomyelitis, and necrotizing fascitis. One of the most useful phenotypic characteristics of streptococci is the hemolytic reaction, generally classified as alpha, beta or gamma according to the appearance of zones around colonies growing on Blood Agar. Beta-hemolytic streptococci are further characterized by Lancefield groups based on the antigenic differences in group-specific polysaccharides located in the bacterial cell wall.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
TRUSTED_SUSTAINABILITY

Specifications

Description Todd Hewitt Broth; LIM Broth; 3mL; 15 x 103mm Tube
Format 15 x 103mm Tube
Manufacturing Origin United States
Quantity 100/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.