Learn More
Teknova Todd Hewitt Broth with 0.5% Yeast Extract. 1 Liter, Sterile. 2 pack.
SDP
Catalog No. 502387576
Encompass
3.00% Bacto Todd-Hewitt Broth ,0.5% Yeast Extract,Bacto is a registered trademark of BD corporation
Catalog No. 50-238-7576 Supplier Teknova Supplier No. T98442PK
3.00% Bacto Todd-Hewitt Broth ,0.5% Yeast Extract,Bacto is a registered trademark of BD corporation