Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Todd Hewitt Broth
GSA_VA

Catalog No. R064805 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100/Pk.
20/Pk.
Format:
15mm x 103mm Tube
Tube
Fill Volume:
5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Format Fill Volume
R064805 20/Pk. 15mm x 103mm Tube 5 mL
R064804 100/Pk. Tube 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. R064805 Supplier Thermo Scientific™ Supplier No. R064805
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Reduce media prep time while cultivating beta hemolytic streptococci and encouraging the production of antigenic streptococci hemolysins with pre-filled, ready-to-use tubes.

Cultivate beta hemolytic streptococci, encourage the production of antigenic streptococci hemolysins, and isolate pathogenic microorganisms using convenient, pre-filled, ready-to-use Thermo Scientific™ Todd Hewitt Broth.

Remove the need for pipetting, dispensing or autoclaving with convenient, pre-filled and ready-to-use tubes of Todd Hewitt Broth, available in multiple formats and formulations.

Streptococci are facultatively anaerobic, catalase-negative, gram-positive cocci that occur as pairs or chains. They cause a wide variety of clinical diseases including pharyngitis, scarlet fever, otitis media, impetigo, meningitis, neonatal sepis, pneumonia, endocarditis, osteomyelitis, and necrotizing fascitis. One of the most useful phenotypic characteristics of streptococci is the hemolytic reaction, generally classified as alpha, beta or gamma according to the appearance of zones around colonies growing on Blood Agar. Beta-hemolytic streptococci are further characterized by Lancefield groups based on the antigenic differences in group-specific polysaccharides located in the bacterial cell wall.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Description Todd Hewitt Broth
Format 15mm x 103mm Tube
Manufacturing Origin United States
Product Type Microbiology Media
Fill Volume 5 mL
Quantity 20/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.