Learn More
Medchemexpress LLC Todralazine hydrochloride | 3778-76-5 | 99.9% | 5 MG
SDP
Supplier:  Medchemexpress LLC HYB1001A5MG
Encompass_Preferred
Todralazine hydrochloride is an anti-hypertensive agent that acts as a β2AR blocker, possessing antioxidant and free radical scavenging activity. It is intended for research use only and not for sale to patients.
  • Anti-hypertensive agent
  • Acts as a β2AR blocker
  • Possesses antioxidant activity
  • Possesses free radical scavenging activity
  • Intended for research use only
Catalog No. 50-004-07507