Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Waters Standard Kit, ToF G2-S, System Performance, Liquid

Catalog No. NC1288538 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

This Standards Kit verifies ToF G2-S system performance, offering a comprehensive set of liquid and powder standards for analytical applications.

  • Includes six distinct standards.
  • Standard 1: Sodium Iodide solution.
  • Standard 2: Sodium Hydroxide solution.
  • Standard 3: PEG Maldi solution.
  • Standard 4: Six-compound mix: Acetaminophen, Caffeine, Sulfadimethoxine, Verapamil, Chloramphenicol, 17-A-Hydroxyprogesterone.
  • Standard 5: Chloramphenicol powder.
  • Standard 6: Sulfadimethoxine powder.
  • MS detection.
  • Liquid formulation.
  • Storage: 2°C to 8°C.
  • LCMS compatible.
  • 6-month shelf life.
  • Ambient shipping.
  • Reference type.
  • 1 unit per package.
  • Hazardous material; special freight handling required.

Catalog No. NC1288538 Supplier Waters Corp Supplier No. 700008892
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This Standards Kit verifies ToF G2-S system performance, offering a comprehensive set of liquid and powder standards for analytical applications.

  • Includes six distinct standards.
  • Standard 1: Sodium Iodide solution.
  • Standard 2: Sodium Hydroxide solution.
  • Standard 3: PEG Maldi solution.
  • Standard 4: Six-compound mix: Acetaminophen, Caffeine, Sulfadimethoxine, Verapamil, Chloramphenicol, 17-A-Hydroxyprogesterone.
  • Standard 5: Chloramphenicol powder.
  • Standard 6: Sulfadimethoxine powder.
  • MS detection.
  • Liquid formulation.
  • Storage: 2°C to 8°C.
  • LCMS compatible.
  • 6-month shelf life.
  • Ambient shipping.
  • Reference type.
  • 1 unit per package.
  • Hazardous material; special freight handling required.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.