Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Waters TOF G2-S Sample Kit, MS, LCMS, System Performance

Catalog No. 502224448 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

This standards kit is designed for the verification of system performance for Tof G2-S. It provides a comprehensive set of standards for various analytical needs.

  • Includes 2 vials of 3 mg Leucine Enkephalin.
  • Includes 2 vials of 0.1 mg [Glu1]-Fibrinopeptide B.
  • Includes 1 vial of 1 nmole Digestion Standard, Alcohol Dehydrogenase.
  • Includes 1 vial of 1 nmole Digestion Standard, Enolase.
  • Detection method: MS.
  • Formulation: Solid.
  • Storage condition: -20°C (freezer).
  • System type: LCMS.
  • Shelf life: 6 months.
  • Shipping condition: Ambient.
  • Type: Reference.
  • Application: System performance.
  • Product type: Standard kits.
  • Units per package: 1.

Catalog No. 50-222-4448 Supplier Waters Corp Supplier No. 700008842
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This standards kit is designed for the verification of system performance for Tof G2-S. It provides a comprehensive set of standards for various analytical needs.

  • Includes 2 vials of 3 mg Leucine Enkephalin.
  • Includes 2 vials of 0.1 mg [Glu1]-Fibrinopeptide B.
  • Includes 1 vial of 1 nmole Digestion Standard, Alcohol Dehydrogenase.
  • Includes 1 vial of 1 nmole Digestion Standard, Enolase.
  • Detection method: MS.
  • Formulation: Solid.
  • Storage condition: -20°C (freezer).
  • System type: LCMS.
  • Shelf life: 6 months.
  • Shipping condition: Ambient.
  • Type: Reference.
  • Application: System performance.
  • Product type: Standard kits.
  • Units per package: 1.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.