Learn More
Medchemexpress LLC HY-101068 100mg Medchemexpress, TOFA RMI14514;MDL14514 CAS:54857-86-2 Purity:98%
SDP
Catalog No. 501636888
Encompass_Preferred
Medchemexpress,HY-101068 100mg TOFA RMI14514;MDL14514 CAS:54857-86-2 Formula:C19H32O4 TOFA (RMI14514;MDL14514) is an allosteric inhibitor of acetyl-CoA carboxylase-α (ACCA ). Purity:98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-163-6888 Supplier Medchemexpress LLC Supplier No. HY101068100MG
Medchemexpress,HY-101068 100mg TOFA RMI14514;MDL14514 CAS:54857-86-2 Formula:C19H32O4 TOFA (RMI14514;MDL14514) is an allosteric inhibitor of acetyl-CoA carboxylase-α (ACCA ). Purity:98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.