Learn More
Selleck Chemical LLC TOFA (5-(Tetradecyloxy)-2-furoic acid) 5mg 54857-86-2 RMI14514, MDL14514
SDP
Supplier:  Selleck Chemical LLC S66905MG
Encompass_Preferred
TOFA (5-(Tetradecyloxy)-2-furoic acid, Tyrphostin 46,Tyrphostin A46,Tyrphostin B40) is an allosteric inhibitor of acetyl-CoA carboxylase-? (ACCA). Inside the cell, TOFA is converted to TOFyl-CoA (5-tetradecyloxy-2-furoyl-CoA), exerting an allosteric inhibition on ACCA. TOFA blocks fatty acid synthesis and induces cell death in a dose-dependent manner. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC1902600