Learn More
Medchemexpress LLC HY-101068 5mg Medchemexpress, TOFA CAS:54857-86-2 Purity:>98%
SDP
Catalog No. 501932025
Encompass_Preferred
Medchemexpress, HY-101068 5mg TOFA CAS:54857-86-2 TOFA (RMI14514;MDL14514) is an allosteric inhibitor of acetyl-CoA carboxylase-α ( ACCA ). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2025 Supplier Medchemexpress LLC Supplier No. HY1010685MG
Medchemexpress, HY-101068 5mg TOFA CAS:54857-86-2 TOFA (RMI14514;MDL14514) is an allosteric inhibitor of acetyl-CoA carboxylase-α ( ACCA ). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.