Learn More
Medchemexpress LLC Tofacitinib 10 mM * 1 mL in DMSO, Solid + Solvent, Ready for Reconstitution
SDP
Catalog No. NC2880848
Encompass_Preferred
Tofacitinib is an orally available JAK3/2/1 inhibitor with IC50s of 1, 20, and 112 nM, respectively.
Catalog No. NC2880848 Supplier Medchemexpress LLC Supplier No. HY4035410MM 1ML
Tofacitinib is an orally available JAK3/2/1 inhibitor with IC50s of 1, 20, and 112 nM, respectively.