Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Tofacitinib-13C3 (Tasocitinib-13C3) | 477600-75-2 | 99.8% | 315.35 | 1 MG
SDP

Catalog No. 5000404070
Encompass_Preferred

Tofacitinib-13C3 is the 13C-labeled Tofacitinib, an orally available JAK3/2/1 inhibitor with IC50s of 1, 20, and 112 nM, respectively.

  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.

Catalog No. 50-004-04070 Supplier Medchemexpress LLC Supplier No. HY40354S1MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Tofacitinib-13C3 is the 13C-labeled Tofacitinib, an orally available JAK3/2/1 inhibitor with IC50s of 1, 20, and 112 nM, respectively.

  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.