Learn More
Medchemexpress LLC HY-40354 200mg Medchemexpress, Tofacitinib CAS:477600-75-2 Purity:>98%
SDP
Catalog No. 502029312
Encompass_Preferred
Medchemexpress, HY-40354 200mg Tofacitinib CAS:477600-75-2 Tofacitinib is an orally available JAK3/2/1 inhibitor with IC50s of 1, 20, and 112 nM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9312 Supplier Medchemexpress LLC Supplier No. HY40354200MG
Medchemexpress, HY-40354 200mg Tofacitinib CAS:477600-75-2 Tofacitinib is an orally available JAK3/2/1 inhibitor with IC50s of 1, 20, and 112 nM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.