Learn More
Medchemexpress LLC HY-40354 500mg Medchemexpress, Tofacitinib Tasocitinib;CP-690550 CAS:477600-75-2 Purity:98%
SDP
Catalog No. 501639000
Encompass_Preferred
Medchemexpress,HY-40354 500mg Tofacitinib Tasocitinib;CP-690550 CAS:477600-75-2 Formula:C16H20N6O Tofacitinib inhibits JAK3 with IC50 of 1 nM while inhibiting JAK2, JAK1, Rock-II and Lck with IC50 values of 20, 112, 3400 and 3870 nM, respectively. Purity:98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-163-9000 Supplier Medchemexpress LLC Supplier No. HY40354500MG
Medchemexpress,HY-40354 500mg Tofacitinib Tasocitinib;CP-690550 CAS:477600-75-2 Formula:C16H20N6O Tofacitinib inhibits JAK3 with IC50 of 1 nM while inhibiting JAK2, JAK1, Rock-II and Lck with IC50 values of 20, 112, 3400 and 3870 nM, respectively. Purity:98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.