Learn More
Medchemexpress LLC Tofacitinib | 477600-75-2 | 99.9% | 312.37 | 100 MG
SDP
Catalog No. 5000364733
Encompass_Preferred
Tofacitinib is an orally available JAK3/2/1 inhibitor with IC50s of 1, 20, and 112 nM, respectively. It is intended for research use only.
  • Inhibits JAK3 with an IC50 of 1 nM
  • Inhibits JAK2 with an IC50 of 20 nM
  • Inhibits JAK1 with an IC50 of 112 nM
Catalog No. 50-003-64733 Supplier Medchemexpress LLC Supplier No. 100MGHY40354
Tofacitinib is an orally available JAK3/2/1 inhibitor with IC50s of 1, 20, and 112 nM, respectively. It is intended for research use only.
  • Inhibits JAK3 with an IC50 of 1 nM
  • Inhibits JAK2 with an IC50 of 20 nM
  • Inhibits JAK1 with an IC50 of 112 nM