Learn More
Medchemexpress LLC Tofacitinib | 477600-75-2 | MFCD22628813 | 99.9% | 10 MG
SDP
Catalog No. 5000297859
Encompass_Preferred
Tofacitinib (Standard) is an analytical reference standard of the JAK inhibitor tofacitinib supplied for research and analytical use. It is provided as a high-purity material suitable for method development, calibration, and quantitative analysis; not for human or clinical use.
  • Analytical reference standard for HPLC and LC-MS method development.
  • High purity suitable for quantitative assay calibration.
  • Supplied in small pack sizes for laboratory testing.
  • Applicable as an in vitro pharmacology reference compound.
  • Not for human or clinical use; intended for research only.
Catalog No. 50-002-97859 Supplier Medchemexpress LLC Supplier No. HY40354R10MG
Tofacitinib (Standard) is an analytical reference standard of the JAK inhibitor tofacitinib supplied for research and analytical use. It is provided as a high-purity material suitable for method development, calibration, and quantitative analysis; not for human or clinical use.
  • Analytical reference standard for HPLC and LC-MS method development.
  • High purity suitable for quantitative assay calibration.
  • Supplied in small pack sizes for laboratory testing.
  • Applicable as an in vitro pharmacology reference compound.
  • Not for human or clinical use; intended for research only.