Learn More
Selleck Chemical LLC Tofacitinib (CP-690550) Citrate S5001-200mg
SDP
Catalog No. 503110187
Encompass_Preferred
Tofacitinib citrate (CP-690550 Tasocitinib) is a novel inhibitor of JAK with IC50 of 1 nM 20 nM and 112 nM against JAK3 JAK2 and JAK1 respectively Tofacitinib citrate has anti-infection activity
Catalog No. 50-311-0187 Supplier Selleck Chemical LLC Supplier No. S5001200MG
Tofacitinib citrate (CP-690550 Tasocitinib) is a novel inhibitor of JAK with IC50 of 1 nM 20 nM and 112 nM against JAK3 JAK2 and JAK1 respectively Tofacitinib citrate has anti-infection activity