Learn More
Medchemexpress LLC HY-40354A 200mg , Tofacitinib (citrate) CAS:540737-29-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY40354A200MG
Encompass_Preferred
HY-40354A 200mg Tofacitinib (citrate) CAS: 540737-29-9 Tofacitinib citrate is an orally available JAK1/2/3 inhibitor with IC50s of 1, 20, and 112 nM, respectively. Tofacitinib citrate has antibacterial, antifungal and antiviral activities. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9273