Learn More
Medchemexpress LLC Tofacitinib citrate | 540737-29-9 | 99.94% | 50 MG
SDP
Catalog No. 5000404072
Encompass_Preferred
Tofacitinib citrate is an orally available JAK3/2/1 inhibitor with IC50s of 1, 20, and 112 nM, respectively. It also exhibits antibacterial, antifungal, and antiviral activities.
  • Orally available JAK3/2/1 inhibitor
  • Antibacterial activity
  • Antifungal activity
  • Antiviral activity
Catalog No. 50-004-04072 Supplier Medchemexpress LLC Supplier No. HY40354A50MG
Tofacitinib citrate is an orally available JAK3/2/1 inhibitor with IC50s of 1, 20, and 112 nM, respectively. It also exhibits antibacterial, antifungal, and antiviral activities.
  • Orally available JAK3/2/1 inhibitor
  • Antibacterial activity
  • Antifungal activity
  • Antiviral activity