Learn More
Selleck Chemical LLC Tofacitinib (CP-690550) Citrate 10mg 540737-29-9
SDP
Supplier:  Selleck Chemical LLC S500110MG
Encompass_Preferred
Tofacitinib citrate (CP-690550 citrate) is a novel inhibitor of JAK with IC50 of 1 nM, 20 nM and 112 nM against JAK3, JAK2, and JAK1, respectively. Tofacitinib citrate has anti-infection activity. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-698-6