Learn More
Medchemexpress LLC Tofacitinib impurity 3 | 1616761-00-2 | 99.84% | C₁₆H₁₉ClN₆O | 5 MG
SDP
Catalog No. 5000434885
Encompass_Preferred
Tofacitinib impurity 3 is an impurity of Tofacitinib, primarily used as a laboratory chemical and for the manufacture of substances.
  • Suitable for laboratory use.
  • Used in the manufacture of substances.
  • Store sealed at 4°C, away from moisture.
  • Stable in solvent at -80°C for 6 months, or -20°C for 1 month.
  • Can be shipped at room temperature if less than 2 weeks.
Catalog No. 50-004-34885 Supplier Medchemexpress LLC Supplier No. HY785605MG
Tofacitinib impurity 3 is an impurity of Tofacitinib, primarily used as a laboratory chemical and for the manufacture of substances.
  • Suitable for laboratory use.
  • Used in the manufacture of substances.
  • Store sealed at 4°C, away from moisture.
  • Stable in solvent at -80°C for 6 months, or -20°C for 1 month.
  • Can be shipped at room temperature if less than 2 weeks.