Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Tofacitinib impurity 4 | 384338-23-2 | 96.0% | 218.34 | 50 MG
SDP

Catalog No. 5000402618
Encompass_Preferred

Tofacitinib impurity 4 is an impurity of Tofacitinib. This product is designed for research use only.

  • Appearance: Liquid
  • Color: Colorless to light yellow
  • Shipping: Room temperature in continental US; may vary elsewhere
  • Storage: Pure form can be stored at -20°C for 3 years or 4°C for 2 years; in solvent, it can be stored at -80°C for 6 months or -20°C for 1 month

Catalog No. 50-004-02618 Supplier Medchemexpress LLC Supplier No. HY2127350MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Tofacitinib impurity 4 is an impurity of Tofacitinib. This product is designed for research use only.

  • Appearance: Liquid
  • Color: Colorless to light yellow
  • Shipping: Room temperature in continental US; may vary elsewhere
  • Storage: Pure form can be stored at -20°C for 3 years or 4°C for 2 years; in solvent, it can be stored at -80°C for 6 months or -20°C for 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.